missing translation for 'onlineSavingsMsg'
Learn More

Methyl geranate, mixture of isomers, 94%

Product Code. 11429496
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429496 10 g 10g
11439496 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429496 Supplier Thermo Scientific Alfa Aesar Supplier No. A19218.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1189-09-9 | C11H18O2 | 182.263 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189-09-9
Molecular Formula C11H18O2
Molecular Weight (g/mol) 182.263
MDL Number MFCD00036571
InChI Key ACOBBFVLNKYODD-CSKARUKUSA-N
Synonym methyl geranate, e-methylgeranate, methyl geraniate, unii-26yr95mgp2, methyl 2e-3,7-dimethylocta-2,6-dienoate, 2,6-octadienoic acid, 3,7-dimethyl-, methyl ester, e, methyl geranate, mixture of isomers, methyl 3,7-dimethyl-2,6-octadienoate, 2,6-octadienoic acid, 3,7-dimethyl-, methyl ester, 2e, 2,6-octadienoic acid, 3,7-dimethyl-, methyl ester
PubChem CID 5365910
IUPAC Name methyl (2E)-3,7-dimethylocta-2,6-dienoate
SMILES CC(=CCCC(=CC(=O)OC)C)C

Specifications

Density 0.925
Boiling Point 70°C (4 mmHg)
Flash Point 99°C (210°F)
Refractive Index 1.469
Quantity 10 g
Formula Weight 182.26
Percent Purity 94%
Chemical Name or Material Methyl geranate, mixture of isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.