missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Green, zinc chloride

Product Code. 15494099
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494099 25 g 25g
15404109 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494099 Supplier Thermo Scientific Chemicals Supplier No. J61509.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7114-03-6 | C27H35Cl4N3Zn | 608.78 g/mol

Methyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.

Solubility
Soluble in water, ethanol or methanol

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7114-03-6
Molecular Formula C27H35Cl4N3Zn
Molecular Weight (g/mol) 608.78
MDL Number MFCD00151094
InChI Key ZKOGXCSOLMZRAZ-UHFFFAOYSA-J
Synonym C.I. 42590
PubChem CID 131675247
IUPAC Name zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride
SMILES CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]

Specifications

Melting Point >300°C
Beilstein 3914106
Color Green
Solubility Information Soluble in water,ethanol or methanol
Quantity 25 g
Physical Form Powder
Formula Weight 653.24
Chemical Name or Material Methyl Green, zinc chloride
compliance-icons
Product Identifier
  • Methyl Green, zinc chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Methyl Green, zinc chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.