missing translation for 'onlineSavingsMsg'
Learn More

Methyl heptadecanoate, Thermo Scientific™

Product Code. 17730649 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17730649 1 g 1g
17740649 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17730649 Supplier Thermo Scientific Acros Supplier No. 469300010
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

Alpha Vector METHYLHEPTADECANOATE
Chemical Name or Material Methyl heptadecanoate
CAS 1731-92-6
Melting Point 29°C to 30°C
Color Colorless or White to Yellow
Boiling Point 152°C to 153°C
Flash Point >110°C
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C18 H36 O2
MDL Number 00009001
Quantity 1 g
Synonym Methyl margarate
Molecular Weight (g/mol) 284.48
Physical Form Liquid or Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.