missing translation for 'onlineSavingsMsg'
Learn More

Methyl hexanoate, 99%, analytical standard for GC

Product Code. 15540544 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15540544 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15540544 Supplier Thermo Scientific Acros Supplier No. 414420050
  • ONLINE EXCLUSIVE PRICE
    103.00 €
    99.70 € / 5g
    Save 3.30 €
    3% Off
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 106-70-7 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Acidity 1mg KOH/g max.
CAS 106-70-7
Melting Point -71.0°C
Color Colorless
Density 0.8840g/mL
Boiling Point 151.0°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H14O2
Linear Formula CH3(CH2)4COOCH3
Refractive Index 1.4047 to 1.4067
MDL Number MFCD00009510
Quantity 5 g
Specific Gravity 0.884
Synonym methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate
Solubility Information Solubility in water: insoluble
InChI Key NUKZAGXMHTUAFE-UHFFFAOYSA-N
SMILES CCCCCC(=O)OC
IUPAC Name methyl hexanoate
Molecular Weight (g/mol) 130.19
PubChem CID 7824
ChEBI CHEBI:77322
Viscosity <30 mPa.s (20°C)
Formula Weight 130.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl hexanoate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl hexanoate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.