missing translation for 'onlineSavingsMsg'
Learn More

Methyl hydrazinocarboxylate, 97%

Product Code. 10655412 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10655412 100 g Glass bottle 100g
10523531 500 g Glass bottle 500g
10569063 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10655412 Supplier Thermo Scientific Acros Supplier No. 166141000
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6294-89-9 | C2H6N2O2 | 90.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6294-89-9
Molecular Formula C2H6N2O2
Molecular Weight (g/mol) 90.08
MDL Number MFCD00007594
InChI Key WFJRIDQGVSJLLH-UHFFFAOYSA-N
Synonym methyl carbazate, methyl hydrazinecarboxylate, methyl hydrazinocarboxylate, methoxycarbohydrazide, methylcarbazate, hydrazinecarboxylic acid, methyl ester, carbomethoxyhydrazide, methyl hydrazinoformate, methoxycarbonyl hydrazine, carbazic acid methyl ester
PubChem CID 80519
IUPAC Name methyl N-aminocarbamate
SMILES COC(=O)NN

Specifications

Melting Point 68°C to 73°C
Color Pink to White
Boiling Point 108°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2NNHCO2CH3
Quantity 100 g
Beilstein 03, I, 46
Fieser 04,333; 07,235
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and benzene,practically insoluble in petroleum ether
Formula Weight 90.08
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Platelets
Chemical Name or Material Methyl hydrazinocarboxylate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl hydrazinocarboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H372-Causes damage to organs.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.