missing translation for 'onlineSavingsMsg'
Learn More

Methyl Indole-2,4,5,6,7-d5-3-acetate, TRC

Product Code. 30479199
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479199 10 mg 10mg
30473837 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30479199 Supplier Toronto Research Chemicals Supplier No. TRCM31347610MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102415-39-4
Molecular Formula C11 D5 H6 N O2
Molecular Weight (g/mol) 194.2414
Synonym 1H-Indole-2,4,5,6,7-d5-3-acetic acid, methyl ester (9CI), 1H-Indole-2,4,5,6,7-d5-3-acetic acid methyl ester, 1H-Indole-3-acetic Acid Methyl Ester-D5 (indole-2,4,5,6,7-D5), Methyl Indole-2,4,5,6,7-d5-3-acetate, 2-(2,4,5,6,7-Pentadeuterio-1H-indol-3-yl)acetic acid methyl ester, 2-(3-Indolyl-2,4,5,6,7-d5)acetic acid methyl ester, Methyl 2-(1H-indol-3-yl-d5)acetate
IUPAC Name methyl 2-(2,4,5,6,7-pentadeuterio-1H-indol-3-yl)acetate
SMILES [2H]c1[nH]c2c([2H])c([2H])c([2H])c([2H])c2c1CC(=O)OC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H11NO2/c1-14-11(13)6-8-7-12-10-5-3-2-4-9(8)10/h2-5,7,12H,6H2,1H3/i2D,3D,4D,5D,7D
Formula Weight 194.1104
Chemical Name or Material Methyl Indole-2,4,5,6,7-d5-3-acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.