missing translation for 'onlineSavingsMsg'
Learn More

Methyl indole-4-carboxylate, 99%, Thermo Scientific™

Product Code. 10133921 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10133921 1 g Glass bottle 1g
10377381 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10133921 Supplier Thermo Scientific Acros Supplier No. 334030010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39830-66-5
MDL Number MFCD00191222
InChI Key WEAXQUBYRSEBJD-UHFFFAOYSA-N
Synonym methyl indole-4-carboxylate, indole-4-carboxylic acid methyl ester, 1h-indole-4-carboxylic acid methyl ester, methyl 4-indolecarboxylate, 4-indole-carboxylic acid methyl ester, 1h-indole-4-carboxylic acid, methyl ester, 4-methoxycarbonylindole, 4-methoxycarbonyl-1h-indole, indole-4-carboxylate, pubchem1705
PubChem CID 2733668
IUPAC Name methyl 1H-indole-4-carboxylate
SMILES COC(=O)C1=C2C=CNC2=CC=C1

Specifications

Melting Point 67.0°C to 71.0°C
Color Green to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H9NO2
Quantity 1 g
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 175.19
Formula Weight 175.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Methyl indole-4-carboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl indole-4-carboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.