missing translation for 'onlineSavingsMsg'
Learn More

Methyl indole-4-carboxylate, 99%

Product Code. 11302867
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302867 1 g 1g
11322867 250 mg 250mg
11312867 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11302867 Supplier Thermo Scientific Alfa Aesar Supplier No. L16021.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 39830-66-5 | C10H9NO2 | 175.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39830-66-5
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00191222
InChI Key WEAXQUBYRSEBJD-UHFFFAOYSA-N
Synonym methyl indole-4-carboxylate, indole-4-carboxylic acid methyl ester, 1h-indole-4-carboxylic acid methyl ester, methyl 4-indolecarboxylate, 4-indole-carboxylic acid methyl ester, 1h-indole-4-carboxylic acid, methyl ester, 4-methoxycarbonylindole, 4-methoxycarbonyl-1h-indole, indole-4-carboxylate, pubchem1705
PubChem CID 2733668
IUPAC Name methyl 1H-indole-4-carboxylate
SMILES COC(=O)C1=C2C=CNC2=CC=C1

Specifications

Melting Point 68°C to 71°C
Quantity 1 g
Beilstein 141826
Formula Weight 175.19
Percent Purity 99%
Chemical Name or Material Methyl indole-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.