missing translation for 'onlineSavingsMsg'
Learn More

Methyl indole-5-carboxylate, 99%, Thermo Scientific™

Product Code. 10567254 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10567254 1 g 1g
10724185 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10567254 Supplier Thermo Scientific Acros Supplier No. 399540010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1011-65-0
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00153023
InChI Key DRYBMFJLYYEOBZ-UHFFFAOYSA-N
Synonym methyl indole-5-carboxylate, indole-5-carboxylic acid methyl ester, 1h-indole-5-carboxylic acid, methyl ester, methylindole-5-carboxylate, 1h-indole-5-carboxylic acid methyl ester, indole5carboxylicacidmethylester, indole-5-carboxylicacidmethylester, indole-5-carboxylate, pubchem7248, 5-methoxycarbonylindole
PubChem CID 2737635
IUPAC Name methyl 1H-indole-5-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)NC=C2

Specifications

Melting Point 126°C to 128°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 175.19
Percent Purity 99%
Chemical Name or Material Methyl indole-5-carboxylate
compliance-icons
Product Identifier
  • Methyl indole-5-carboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.