missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Indole-5-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | 1H-Indole-5-carboxylic Acid Methyl Ester |
| CAS | 1011-65-0 |
| Molecular Formula | C10 H9 N O2 |
| Quantity | 5 g |
| Synonym | 1H-Indole-5-carboxylic acid','methyl ester, Indole-5-carboxylic acid','methyl ester (6CI,7CI,8CI), 5-(Methoxycarbonyl)-1H-indole, 5-(Methoxycarbonyl)indole, Methyl 1H-indole-5-carboxylate, Methyl 5-indolecarboxylate, 5-(Methoxycarbonyl)indole, H-Indole-5-carboxylic Acid Methyl Ester, 5-(Methoxycarbonyl)-1H-indole |
| InChI Formula | InChI=1S/C10H9NO2/c1-13-10(12)8-2-3-9-7(6-8)4-5-11-9/h2-6,11H,1H3 |
| SMILES | COC(=O)c1ccc2[nH]ccc2c1 |
| IUPAC Name | methyl 1H-indole-5-carboxylate |
| Molecular Weight (g/mol) | 175.184 |
| Formula Weight | 175.0633 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?