Learn More
Methyl isonicotinate, 98%
Description
Methyl Isonicotinate induces increased walking and take-off behavior in western flower thrips, Frankliniella occidentalis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl Isonicotinate induces increased walking and take-off behavior in western flower thrips, Frankliniella occidentalis.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 8°C to 8.5°C |
| Density | 1.016 |
| Boiling Point | 207°C to 209°C |
| Flash Point | 82°C (180°F) |
| Odor | Characteristic |
| Refractive Index | 1.5135 |
| Quantity | 100 g |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 137.14 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
- Methyl isonicotinate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: 4-Pyridinecarboxylicacid, methyl ester
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.