missing translation for 'onlineSavingsMsg'
Learn More

Methyl Isonicotinate, TRC

Product Code. 30478005
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478005 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478005 Supplier Toronto Research Chemicals Supplier No. TRCM323340100G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Isonicotinate
CAS 2459-09-8
Molecular Formula C7H7NO2
Quantity 100 g
Synonym 4-Pyridinecarboxylic Acid Methyl Ester, Isonicotinic Acid Methyl Ester, 4-Carbomethoxypyridine, 4-Methoxycarbonylpyridine, Methyl 4-Pyridinecarboxylate, NSC 18257
InChI Formula InChI=1S/C7H7NO2/c1-10-7(9)6-2-4-8-5-3-6/h2-5H,1H3
SMILES COC(=O)c1ccncc1
IUPAC Name methyl pyridine-4-carboxylate
Molecular Weight (g/mol) 137.14
Formula Weight 137.0477
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.