missing translation for 'onlineSavingsMsg'
Learn More

Methyl isonipecotate, 97%, Thermo Scientific™

Product Code. 10295144 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10295144 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10295144 Supplier Thermo Scientific Acros Supplier No. 398751000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2971-79-1
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00190578
InChI Key RZVWBASHHLFBJF-UHFFFAOYSA-N
Synonym methyl isonipecotate, methyl 4-piperidinecarboxylate, 4-piperidinecarboxylic acid, methyl ester, piperidine-4-carboxylic acid methyl ester, 4-piperidinecarboxylic acid methyl ester, isonipecotic acid methyl ester, methylpiperidine-4-carboxylate, methylisonipecotate, methyl piperidin-4-carboxylate, acmc-20aiv4
PubChem CID 424914
IUPAC Name methyl piperidine-4-carboxylate
SMILES COC(=O)C1CCNCC1

Specifications

Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 85.0°C to 90.0°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4640 to 1.4660 (20°C, 589nm)
Quantity 100 mL
Formula Weight 143.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl isonipecotate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl isonipecotate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.