missing translation for 'onlineSavingsMsg'
Learn More

Methyl isoxazole-5-carboxylate, 97%

Product Code. 11450618
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450618 5 g 5g
11440618 1 g 1g
11460618 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450618 Supplier Thermo Scientific Alfa Aesar Supplier No. B25169.06
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15055-81-9 | C5H5NO3 | 127.099 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15055-81-9
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
MDL Number MFCD01570794
InChI Key ILPCPKZAAQXHKL-UHFFFAOYSA-N
Synonym methyl isoxazole-5-carboxylate, 5-isoxazolecarboxylic acid, methyl ester, isoxazole-5-carboxylic acid methyl ester, acmc-209wih, methyl 5-isoxazolcarboxylate, methyl 5-isoxazolcarboxylate;, methylisoxazole-5-carboxylate, 5-methoxycarbonyl-1,2-oxazol, 5-isoxazol-carbonsaeure-methylester;, isoxazol-5-carbonsaeure-methylester;
PubChem CID 2736892
IUPAC Name methyl 1,2-oxazole-5-carboxylate
SMILES COC(=O)C1=CC=NO1

Specifications

Melting Point 48°C to 50°C
Quantity 5 g
Formula Weight 127.1
Percent Purity 97%
Chemical Name or Material Methyl isoxazole-5-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.