missing translation for 'onlineSavingsMsg'
Learn More

Methyl laurate, 98+%

Product Code. 10540604 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10540604 50 g Glass bottle 50g
10762831 2.5 g Glass bottle 2.5g
10127663 10 g Glass bottle 10g
15149233 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10540604 Supplier Thermo Scientific Acros Supplier No. 215560500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 111-82-0 | C13H26O2 | 214.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-82-0
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.35
MDL Number MFCD00008966
InChI Key UQDUPQYQJKYHQI-UHFFFAOYSA-N
Synonym methyl laurate, lauric acid methyl ester, dodecanoic acid, methyl ester, methyl laurinate, methyl dodecylate, methyl n-dodecanoate, uniphat a40, metholene 2296, lauric acid, methyl ester, stepan c40
PubChem CID 8139
ChEBI CHEBI:87494
IUPAC Name methyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC

Specifications

Melting Point 4°C to 5°C
Color Colorless to Yellow
Density 0.8700g/mL
Boiling Point 261°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)10CO2CH3
Refractive Index 1.4310 to 1.4330
Quantity 50 g
Beilstein 02,361
Specific Gravity 0.87
Solubility Information Solubility in water: insoluble
Formula Weight 214.35
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Methyl laurate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl laurate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Dodecanoic acid, methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.