missing translation for 'onlineSavingsMsg'
Learn More

Methyl Laurate, TRC

Product Code. 30012558
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012558 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012558 Supplier Toronto Research Chemicals Supplier No. TRCM3156502.5G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 111-82-0
Molecular Formula C13 H26 O2
Quantity 2.5 g
Synonym Dodecanoic acid, methyl ester, Lauric acid, methyl ester (6CI,8CI), Agnique ME 1270U, Agnique ME 1290, Agnique ME 1298, CE 1290, CE 1295, Edenor ME-C 1298-100, Emery 2296, Exceparl ML 85, Metholene 2296, Methyl dodecanoate, Methyl dodecylate, Methyl laurate, Methyl laurinate, Methyl n-dodecanoate, NSC 5027, Pastell M 12, Radia 7118, Stepan C 40, Texaprint SDM 100, Uniphat A 40
InChI Formula InChI=1S/C13H26O2/c1-3-4-5-6-7-8-9-10-11-12-13(14)15-2/h3-12H2,1-2H3
SMILES CCCCCCCCCCCC(=O)OC
IUPAC Name methyl dodecanoate
Molecular Weight (g/mol) 214.34
Formula Weight 214.1933
Percent Purity >95%
Chemical Name or Material Dodecanoic acid-methyl ester
Grade GC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.