missing translation for 'onlineSavingsMsg'
Learn More

Methyl linoleate, 95%

Product Code. p-7044619
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11486740

Brand: Thermo Scientific Alfa Aesar L07450.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-63-0 | C19H34O2 | 294.479 g/mol

The oxidation of methyl linoleate in organic solution and phosphatidylcholine (PC) in liposomal membranes induced by an azo initiator has often been used as a model for lipid peroxidation. Methyl linoleate may be used as a model compound in oxidation/peroxidation assays to evaluate the anti-peroxidation activity of fullerenes, ellagitannins and other natural products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The oxidation of methyl linoleate in organic solution and phosphatidylcholine (PC) in liposomal membranes induced by an azo initiator has often been used as a model for lipid peroxidation. Methyl linoleate may be used as a model compound in oxidation/peroxidation assays to evaluate the anti-peroxidation activity of fullerenes, ellagitannins and other natural products.

Solubility
Soluble in DMF, fat solvents, oils.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-63-0
Molecular Formula C19H34O2
Molecular Weight (g/mol) 294.479
MDL Number MFCD00009534
InChI Key WTTJVINHCBCLGX-NQLNTKRDSA-N
Synonym methyl linoleate, linoleic acid methyl ester, methyl lineoleate, methyl 9z,12z-octadeca-9,12-dienoate, methyl octadecadienoate, methyl 9-cis,12-cis-octadecadienoate, linoleic acid,methyl ester, linoleic acid, methyl ester, methyl linoleate, native, 9,12-octadecadienoic acid z,z-, methyl ester
PubChem CID 5284421
ChEBI CHEBI:69080
IUPAC Name methyl (9Z,12Z)-octadeca-9,12-dienoate
SMILES CCCCCC=CCC=CCCCCCCCC(=O)OC

Specifications

Melting Point -35°C
Density 0.888
Boiling Point 207°C to 208°C (11 mmHg)
Flash Point 200°C (392°F)
Refractive Index 1.464
Quantity 1 g
Beilstein 1727614
Merck Index 14,6095
Solubility Information Soluble in DMF,fat solvents,oils.
Formula Weight 294.47
Percent Purity 95%
Chemical Name or Material Methyl linoleate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.