missing translation for 'onlineSavingsMsg'
Learn More

Methyl Linoleate, TRC

Product Code. 30133804
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133804 10 g 10g
30136666 50 g 50g
30010699 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133804 Supplier Toronto Research Chemicals Supplier No. TRCM26519010G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-63-0
Molecular Formula C19 H34 O2
Molecular Weight (g/mol) 294.47
Synonym 9,12-Octadecadienoic acid (Z,Z)-, methyl ester, Linoleic acid, methyl ester (6CI,8CI), (9Z,12Z)-9,12-Octadecadienoic acid methyl ester, (9Z,12Z)-Octadecadienoic acid methyl ester, (Z,Z)-9,12-Octadecadienoic acid methyl ester, Methyl (9Z,12Z)-octadeca-9,12-dienoate, Methyl (9Z,12Z)-octadecadienoate, Methyl (Z,Z)-9,12-octadecadienoate, Methyl 9-cis,12-cis-octadecadienoate, Methyl cis,cis-9,12-octadecadienoate, Methyl cis-9,cis-12 linoleate, Methyl cis-9,cis-12-octadecadienoate, Methyl linoleate, Methyl octadec-9,12-dienoate, Radia 7062
IUPAC Name methyl (9Z,12Z)-octadeca-9,12-dienoate
SMILES CCCCC\C=C/C\C=C/CCCCCCCC(=O)OC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C19H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h7-8,10-11H,3-6,9,12-18H2,1-2H3/b8-7-,11-10-
Formula Weight 294.2559
Chemical Name or Material Linoleic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.