missing translation for 'onlineSavingsMsg'
Learn More

Methyl linolenate, MedChemExpress

Product Code. 16490478
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
1 g
5 g
Unit Size:
1g
200mg
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490478 5 g 5g
16471909 1 g 1g
16461906 500 mg 500mg
16431306 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490478 Supplier Medchem Express Supplier No. HY21268
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Methyl linolenate is a polyunsaturated fattly acid (PUFA).

Methyl linolenate is a polyunsaturated fattly acid (PUFA).

Specifications

Chemical Name or Material Methyl linolenate
CAS 301-00-8
Color Colorless
Purity Grade Notes Research
Molecular Formula C19H32O2
Quantity 5 g
Synonym Linolenic acid methyl ester
Solubility Information DMSO : 100 mg/mL (341.93 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC/C=C\C/C=C\C/C=C\CCCCCCCC(OC)=O
Molecular Weight (g/mol) 292.46
Formula Weight 292.46
Percent Purity 98.0%
Grade Research
Physical Form Liquid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.