missing translation for 'onlineSavingsMsg'
Learn More

Methyl methacrylate, 99%, stabilized

Product Code. 10071140 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
25mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10071140 1 L Glass bottle 1 L
10562521 25 mL Glass bottle 25mL
10153510 2.5 L Glass bottle 2.5 L
10791964 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10071140 Supplier Thermo Scientific Acros Supplier No. 127140010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Methyl methacrylate, 99%, C5H8O2, CAS Number-80-62-6, 150-76-5, acryester m, diakon, methacrylic acid methyl ester, methyl 2-methyl-2-propenoate, methyl 2-methylpropenoate, methyl-methacrylat, methylmethacrylate, methyl methacrylate, methyl methylacrylate, pegalan, 1L | CAS: 80-62-6 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-62-6 , 150-76-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008587
InChI Key VVQNEPGJFQJSBK-UHFFFAOYSA-N
Synonym methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
PubChem CID 6658
ChEBI CHEBI:34840
IUPAC Name methyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC

Specifications

Acidity 0.005 % max. (as methacrylic acid)
Melting Point -48.0°C
Color White to Yellow
Density 0.9300g/mL
Boiling Point 100.0°C
Flash Point 8°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2CH3
Refractive Index 1.4130 to 1.4150
Quantity 1 L
Beilstein 02, II, 398
Merck Index 15, 6013
Specific Gravity 0.93
Solubility Information Solubility in water: 15.9g/L (20°C). Other solubilities: soluble in most common organic solvents such as me,tetrahydrofuran,esters,aromatic and chlorinated,ethanol,ether,acetone
Stabilizer 10 to 110ppm MEHQ
Viscosity 0.6 mPa.s (20°C)
Formula Weight 100.12
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl methacrylate, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl methacrylate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.