missing translation for 'onlineSavingsMsg'
Learn More

Methyl methacrylate, 99%, stabilized

Product Code. 10562521 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
25mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10562521 25 mL Glass bottle 25mL
10071140 1 L Glass bottle 1 L
10153510 2.5 L Glass bottle 2.5 L
10791964 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10562521 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 127140250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Methyl methacrylate, 99%, C5H8O2, CAS Number-80-62-6, 150-76-5, acryester m, diakon, methacrylic acid methyl ester, methyl 2-methyl-2-propenoate, methyl 2-methylpropenoate, methyl-methacrylat, methylmethacrylate, methyl methacrylate, methyl methylacrylate, pegalan, 1L | CAS: 80-62-6 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 80-62-6 , 150-76-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008587
InChI Key VVQNEPGJFQJSBK-UHFFFAOYSA-N
Synonym methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
PubChem CID 6658
ChEBI CHEBI:34840
IUPAC Name methyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC

Especificaciones

Acidity 0.005 % max. (as methacrylic acid)
Melting Point -48.0°C
Color White to Yellow
Density 0.9300g/mL
Boiling Point 100.0°C
Flash Point 8°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2CH3
Refractive Index 1.4130 to 1.4150
Quantity 25 mL
Beilstein 02, II, 398
Merck Index 15, 6013
Specific Gravity 0.93
Solubility Information Solubility in water: 15.9g/L (20°C). Other solubilities: soluble in most common organic solvents such as me,tetrahydrofuran,esters,aromatic and chlorinated,ethanol,ether,acetone
Stabilizer 10 to 110ppm MEHQ
Viscosity 0.6 mPa.s (20°C)
Formula Weight 100.12
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl methacrylate, Stabilized
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • Methyl methacrylate
Palabra de advertencia
  • Danger
Clasificación
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.