missing translation for 'onlineSavingsMsg'
Learn More

Methyl Methacrylate, Extra Pure, SLR, Stabilised with Topanol, Fisher Chemical™

Product Code. 10569000 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10569000 500 mL 500mL
10459370 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10569000 Supplier Fisher Chemical Supplier No. M/4950/08
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 80-62-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number 8587
InChI Key VVQNEPGJFQJSBK-UHFFFAOYSA-N
Synonym methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
PubChem CID 6658
ChEBI CHEBI:34840
IUPAC Name methyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC

Specifications

Melting Point -48°C
Color Colorless
Boiling Point 100°C
Packaging Amber glass bottle
Quantity 500 mL
Vapor Density 3.5
Formula Weight 100.12g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Methyl methacrylate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.