missing translation for 'onlineSavingsMsg'
Learn More

Methyl Methacrylate, TRC

Product Code. 30005388
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005388 10 g 10g
30011226 50 g 50g
17975095 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005388 Supplier Toronto Research Chemicals Supplier No. TRCM32130010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-62-6
Molecular Formula C5 H8 O2
Molecular Weight (g/mol) 100.12
Synonym Methacrylic acid methyl ester (6CI,8CI), Methacrylic acid, methyl ester (7CI), 2-(Methoxycarbonyl)-1-propene, 2-Methyl-2-propenoic acid methyl ester, ACR-PA 20, Acryester M, Acryester MMA, Acryloid HT 100, DL 101, Light Ester M, MMA, Methyl 2-methacrylate, Methyl 2-methyl-2-propenoate, Methyl 2-methylacrylate, Methyl isobutenoate, Methyl methacrylate, NSC 4769, Paraloid HT 100, Pegalan, TEB 3K, Three bond 3057J
IUPAC Name methyl 2-methylprop-2-enoate
SMILES COC(=O)C(=C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H8O2/c1-4(2)5(6)7-3/h1H2,2-3H3
Formula Weight 100.0524
Percent Purity >95%
Chemical Name or Material Methacrylic acid-methyl ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.