missing translation for 'onlineSavingsMsg'
Learn More

Methyl Methanethiosulfonate, TRC

Product Code. 30137117
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137117 1 g 1g
30135879 5 g 5g
17901845 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137117 Supplier Toronto Research Chemicals Supplier No. TRCM3215001G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2949-92-0
Molecular Formula C2 H6 O2 S2
Molecular Weight (g/mol) 126.2
Synonym S-Methyl Methanethiosulfonate
IUPAC Name methylsulfonylsulfanylmethane
SMILES CSS(=O)(=O)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2H6O2S2/c1-5-6(2,3)4/h1-2H3
Formula Weight 125.9809
Chemical Name or Material Methyl Methanethiosulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.