missing translation for 'onlineSavingsMsg'
Learn More

Methyl (methylthio)methyl sulfoxide, 97%

Product Code. 15468779
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468779 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15468779 Supplier Thermo Scientific Alfa Aesar Supplier No. L00510.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33577-16-1 | C3H8OS2 | 124.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 33577-16-1
Density 1.222
Boiling Point 222°C to 226°C
Flash Point >110°C (230°F)
Molecular Formula C3H8OS2
Refractive Index 1.552
MDL Number MFCD00002091
Quantity 10 g
Beilstein 906789
Sensitivity Hygroscopic
Synonym methyl methylsulfinylmethyl sulfide, methyl methylsulfinyl methyl sulfide, methane, methylsulfinyl methylthio, methyl methylthio methyl sulfoxide, famso, methanesulfinyl-methylsulfanyl-methane, formaldehyde dimethyl mercaptal s-oxide, methylsulfinyl methylthio methane, methyl methylthiomethyl sulphoxide, formaldehyde methyl mercaptal s-oxide
InChI Key OTKFCIVOVKCFHR-UHFFFAOYNA-N
SMILES CSCS(C)=O
IUPAC Name methylsulfanyl(methylsulfinyl)methane
Molecular Weight (g/mol) 124.22
PubChem CID 99129
Formula Weight 124.22
Percent Purity 97%
Chemical Name or Material Methyl (methylthio)methyl sulfoxide
Show More Show Less
compliance-icons
Product Identifier
  • Methyl (methylthio)methyl sulfoxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Methyl (methylthio)methyl sulfoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.