missing translation for 'onlineSavingsMsg'
Learn More

Methyl myristate, 99%, analytical standard for GC

Product Code. 15570634 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15570634 5 g 5g
15560634 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15570634 Supplier Thermo Scientific Acros Supplier No. 460360050
  • ONLINE EXCLUSIVE PRICE
    103.00 €
    100.00 € / 5g
    Save 3.00 €
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124-10-7 | C15H30O2 | 242.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-10-7
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.40
MDL Number MFCD00008983
InChI Key ZAZKJZBWRNNLDS-UHFFFAOYSA-N
Synonym methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci
PubChem CID 31284
IUPAC Name methyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 18.4°C
Density 0.8630g/mL
Boiling Point 323.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)12CO2CH3
Refractive Index 1.4340 to 1.4380
Quantity 5 g
Specific Gravity 0.863
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol
Formula Weight 242.4
Percent Purity 99%
Chemical Name or Material Methyl myristate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.