missing translation for 'onlineSavingsMsg'
Learn More

Methyl nicotinate, 99%

Product Code. 11466766
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466766 500 g 500g
11456766 100 g 100g
11428800 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11466766 Supplier Thermo Scientific Alfa Aesar Supplier No. B23908.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93-60-7 | C7H7NO2 | 137.138 g/mol

Used as a rubefacient, topical vasodilator used to test the efficacy of skin protection compounds (i.e. products designed to provide barriers to chemical warfare agents) and used as a flavoring agent. It is a useful synthetic intermediate. It can be used to prepare biarylcarboxamide inhibitors of phosphodiesterase IV and tumor necrosis factor-α in treatment of rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a rubefacient, topical vasodilator used to test the efficacy of skin protection compounds (i.e. products designed to provide barriers to chemical warfare agents) and used as a flavoring agent. It is a useful synthetic intermediate. It can be used to prepare biarylcarboxamide inhibitors of phosphodiesterase IV and tumor necrosis factor-α in treatment of rheumatoid arthritis.

Solubility
Soluble in water at 20°C 48 g/L. Soluble in alcohol, chloroform and ethyl acetate. Solubility in methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-60-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00006388
InChI Key YNBADRVTZLEFNH-UHFFFAOYSA-N
Synonym methyl nicotinate, nikomet, methylnicotinate, nicotinic acid methyl ester, nicometh, methyl 3-pyridinecarboxylate, methyl-nicotinate, 3-pyridinecarboxylic acid, methyl ester, heat spray, nicotinic acid, methyl ester
PubChem CID 7151
IUPAC Name methyl pyridine-3-carboxylate
SMILES COC(=O)C1=CN=CC=C1

Specifications

Melting Point 40°C to 44°C
Boiling Point 208°C to 210°C
Flash Point 95°C (203°F)
Odor Slightly Sweetish
Quantity 500 g
Beilstein 113951
Merck Index 14,6100
Solubility Information Soluble in water at 20°C 48g/L. Soluble in alcohol,chloroform and ethyl acetate. Solubility in methanol (almost transparency).
Formula Weight 137.14
Percent Purity 99%
Chemical Name or Material Methyl nicotinate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl nicotinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Methyl nicotinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.