missing translation for 'onlineSavingsMsg'
Learn More

Methyl nicotinoylacetate, 95%

Product Code. 10147950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10147950 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10147950 Supplier Thermo Scientific Acros Supplier No. 425490025
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 54950-20-8 | C9H9NO3 | 179.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 54950-20-8
Melting Point 34.0°C to 42.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C9H9NO3
MDL Number MFCD00216524
Quantity 2.5 g
Synonym methyl nicotinoylacetate, methyl 3-oxo-3-pyridin-3-yl propanoate, 3-oxo-3-pyridin-3-ylpropionic acid methyl ester, 3-pyridinepropanoic acid, .beta.-oxo-, methyl ester, methyl 3-oxo-3-3-pyridyl propanoate, methyl 3-pyridinecarbonyl acetate, methyl 2-nicotinoyl acetate, 3-pyridinepropanoic acid, beta-oxo-, methyl ester, methyl-3-oxo-3 3-pyridinyl propanoate
Solubility Information Solubility in water: neglegible
InChI Key JUQKVXRLRKKRPL-UHFFFAOYSA-N
SMILES COC(=O)CC(=O)C1=CC=CN=C1
IUPAC Name methyl 3-oxo-3-pyridin-3-ylpropanoate
Molecular Weight (g/mol) 179.18
PubChem CID 108645
Formula Weight 179.17
Percent Purity 95%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Methyl nicotinoylacetate, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.