missing translation for 'onlineSavingsMsg'
Learn More

Methyl non-2-ynoate, ≥95%, Maybridge™

Product Code. 10150742
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10150742

Brand: Thermo Scientific Maybridge SB00926EE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 111-80-8
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.236
MDL Number MFCD00009547
InChI Key NTLJTUMJJWVCTL-UHFFFAOYSA-N
Synonym methyl 2-nonynoate, 2-nonynoic acid methyl ester, methyl octine carbonate, methyl octyne carbonate, 2-nonynoic acid, methyl ester, methyloctyne carboxylate, methyloctyne carbonate, methyl octin carbonate, octynecarboxylic acid, methyl ester, methyl octynecarboxylate
PubChem CID 8137
ChEBI CHEBI:51749
IUPAC Name methyl non-2-ynoate
SMILES CCCCCCC#CC(=O)OC

Specifications

CAS Min % 95.0
Assay Percent Range 95%+
Linear Formula CH3(CH2)5Cidentical toCC(O)OCH3
Quantity 50 g
Formula Weight 168.23
Percent Purity 95%
Chemical Name or Material Methyl non-2-ynoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.