missing translation for 'onlineSavingsMsg'
Learn More

Methyl Octanoate, TRC

Product Code. 30004613
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004613 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30004613 Supplier Toronto Research Chemicals Supplier No. TRCM32449010G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 111-11-5
Molecular Formula C9 H18 O2
Quantity 10 g
Synonym Octanoic Acid Methyl Ester, Octanoic acid, methyl ester, Agnique ME 890, Caprylic acid methyl ester, Methyl caprylate, Methyl n-octanoate, Methyl octanoate, NSC 3710, Pastell M 08, Pastell M 8, Radia 7984, Uniphat A 20, Witconol 1095, Witconol 2309
InChI Formula InChI=1S/C9H18O2/c1-3-4-5-6-7-8-9(10)11-2/h3-8H2,1-2H3
SMILES CCCCCCCC(=O)OC
IUPAC Name methyl octanoate
Molecular Weight (g/mol) 158.24
Formula Weight 158.1307
Chemical Name or Material Caprylic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.