missing translation for 'onlineSavingsMsg'
Learn More

Methyl oleate, 99%, analytical standard for GC

Product Code. 15500644 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15500644 1 g 1g
15510644 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15500644 Supplier Thermo Scientific Acros Supplier No. 460380010
  • ONLINE EXCLUSIVE PRICE
    37.70 €
    36.60 € / 1g
    Save 1.10 €
    2% Off
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 112-62-9 | C19H36O2 | 296.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-62-9
Molecular Formula C19H36O2
Molecular Weight (g/mol) 296.48
InChI Key QYDYPVFESGNLHU-KHPPLWFESA-N
Synonym methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112
PubChem CID 5364509
ChEBI CHEBI:27542
IUPAC Name methyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OC

Specifications

Melting Point -20.0°C
Density 0.8700g/mL
Boiling Point 186.0°C (9.0 mmHg)
Flash Point 180°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)7CHCH(CH2)7COOCH3
Refractive Index 1.4510 to 1.4580
Quantity 1 g
Specific Gravity 0.87
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Viscosity 6.829 mPa.s (20°C)
Formula Weight 296.48
Percent Purity 99%
Chemical Name or Material Methyl oleate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.