missing translation for 'onlineSavingsMsg'
Learn More

Methyl Oleate, TRC

Product Code. 30477903
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477903 25 g 25g
30477153 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477903 Supplier Toronto Research Chemicals Supplier No. TRCM33011025G
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-62-9
Molecular Formula C19 H36 O2
Molecular Weight (g/mol) 296.49
Synonym 9-Octadecenoic acid, methyl ester, Z)-, Oleic acid, methyl ester (6CI,8CI), 9(Z)-Octadecenoic acid methyl ester, Methyl 9(Z)-octadecenoate, Methyl oleate, Methyl cis-9-octadecenoate, cis-Oleic acid methyl ester, Oleic Acid Methyl Ester, 9-Octadecenoic acid, methyl ester, (9Z)-
IUPAC Name methyl (Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3/b11-10-
Formula Weight 296.2715
Chemical Name or Material Oleic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.