missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange Indicator, Reag. Ph. Eur., Solstice

Product Code. 15653820 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
250 g
1.5 kg
Packaging:
Glass Bottle
Unit Size:
1.5kg
100g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15653820 100 g Glass Bottle 100g
15663820 250 g Glass Bottle 250g
15643820 1.5 kg Glass Bottle 1.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15653820 Supplier Honeywell-Fluka Supplier No. 32624100G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Indicator, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
MDL Number MFCD00007502
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
Synonym Helianthin
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

UN Number UN3143
Beilstein 4732884
Formula Weight 327.33g/mol
Packaging Glass Bottle
Suitability Complies for sensitivity
Visual Transition Interval 3.0 to 4.4
Color Orange
pH 3.0 to 4.4 (pink to yellow)
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material Methyl Orange, Reag. Ph. Eur.
Show More Show Less
compliance-icons
Product Identifier
  • Methyl Orange
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium 4-(4- dimethylaminophenylazo) benzenesulphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.