missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl orotate, 97%

Product Code. 15401519
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15401519

Brand: Alfa Aesar™ H64351.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6153-44-2
Molecular Formula C6H6N2O4
Molecular Weight (g/mol) 170.12
MDL Number MFCD00010564
InChI Key UUTDWTOZAWFKFW-UHFFFAOYSA-N
Synonym methyl orotate, orotic acid methyl ester, methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate, unii-41c6ifg4mq, 4-pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester, 6-carbomethyoxyuracil, 41c6ifg4mq, methyl 2,6-dihydroxypyrimidine-4-carboxylate, methyl 2,6-dihydroxy-4-pyrimidinecarboxylate, methyl 2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate
PubChem CID 80257
IUPAC Name methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
SMILES COC(=O)C1=CC(=O)NC(=O)N1

Specifications

Melting Point 240°C to 244°C
Color Yellow
Quantity 1 g
Beilstein 881031
Merck Index 14,6876
Formula Weight 170.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Methyl orotate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.