missing translation for 'onlineSavingsMsg'
Learn More

Methyl orotate, 95%

Product Code. 10444145 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10444145 100 g Glass bottle 100g
10104840 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10444145 Supplier Thermo Scientific Acros Supplier No. 208781000
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 6153-44-2 | C6H6N2O4 | 170.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-44-2
Molecular Formula C6H6N2O4
Molecular Weight (g/mol) 170.12
MDL Number MFCD00010564
InChI Key UUTDWTOZAWFKFW-UHFFFAOYSA-N
Synonym methyl orotate, orotic acid methyl ester, methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate, unii-41c6ifg4mq, 4-pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester, 6-carbomethyoxyuracil, 41c6ifg4mq, methyl 2,6-dihydroxypyrimidine-4-carboxylate, methyl 2,6-dihydroxy-4-pyrimidinecarboxylate, methyl 2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate
PubChem CID 80257
IUPAC Name methyl 2,4-dioxo-1H-pyrimidine-6-carboxylate
SMILES COC(=O)C1=CC(=O)NC(=O)N1

Specifications

Melting Point 244°C
Color Brown or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 100 g
Beilstein 25, 254
Merck Index 15, 6972
Solubility Information Solubility in water: .. Other solubilities: soluble in 1n nh4oh
Formula Weight 170.12
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Methyl orotate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.