missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-tolyl sulfoxide, 98%

Product Code. 10403350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10403350 5 g Glass bottle 5g
10575290 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10403350 Supplier Thermo Scientific Acros Supplier No. 428150050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 934-72-5 | C8H10OS | 154.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 934-72-5
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.23
MDL Number MFCD00075175
InChI Key FEVALTJSQBFLEU-UHFFFAOYNA-N
Synonym 1-methyl-4-methylsulfinyl benzene, methyl 4-methylphenylsulfoxide, methyl p-tolyl sulfoxide, 1-methanesulfinyl-4-methylbenzene, methyl p-methylphenylsulfoxide, 1-methyl-4-methylsulphinyl benzene, benzene, 1-methyl-4-methylsulfinyl, acmc-20aotg, acmc-20apjq, methyl p-tolylsulfoxide
PubChem CID 136743
IUPAC Name 1-methyl-4-methylsulfinylbenzene
SMILES CC1=CC=C(C=C1)S(C)=O

Specifications

Melting Point 44°C to 46°C
Boiling Point 113°C to 114°C (2.2 mmHg)
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 154.23
Percent Purity 98%
Chemical Name or Material Methyl p-tolyl sulfoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.