missing translation for 'onlineSavingsMsg'
Learn More

Methyl palmitate, 97%

Product Code. 11307475
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11307475

Brand: Thermo Scientific Alfa Aesar L05509.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-39-0 | C17H34O2 | 270.457 g/mol

Methyl palmitate is used in the preparation of detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and animal feeds. It exhibits an anti-inflammatory and anti-fibrotic agent and prevents bleomycin-induced lung inflammation and fibrosis in rats. In addition, it also prevents carbon tetrachloride-induced liver fibrosis linked to reduce transforming growth factor beta, which is a secreted protein that controls proliferation, cellular differentiation and other functions in most cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl palmitate is used in the preparation of detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and animal feeds. It exhibits an anti-inflammatory and anti-fibrotic agent and prevents bleomycin-induced lung inflammation and fibrosis in rats. In addition, it also prevents carbon tetrachloride-induced liver fibrosis linked to reduce transforming growth factor beta, which is a secreted protein that controls proliferation, cellular differentiation and other functions in most cells.

Solubility
Soluble in ethanol, chloroform, dimethyl sulfoxide and dimethyl formamide. Insoluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-39-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00008994
InChI Key FLIACVVOZYBSBS-UHFFFAOYSA-N
Synonym methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i
PubChem CID 8181
ChEBI CHEBI:69187
IUPAC Name methyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 28°C to 34°C
Density 0.852
Boiling Point 135°C to 137°C (0.2 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3(CH2)14CO2CH3
Refractive Index 1.4512
Quantity 100 g
Beilstein 1780973
Solubility Information Soluble in ethanol,chloroform,dimethyl sulfoxide and dimethyl formamide. Insoluble in water.
Formula Weight 270.46
Percent Purity 97%
Chemical Name or Material Methyl palmitate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.