missing translation for 'onlineSavingsMsg'
Learn More

Methyl pentafluoropropionate, 98%, Thermo Scientific™

Product Code. 11305149
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305149 5 g 5g
11315149 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11305149 Supplier Thermo Scientific Alfa Aesar Supplier No. A17887.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 378-75-6
Molecular Formula C4H3F5O2
Molecular Weight (g/mol) 178.058
MDL Number MFCD00000430
InChI Key JMKJCPUVEMZGEC-UHFFFAOYSA-N
Synonym methyl pentafluoropropionate, methyl perfluoropropionate, methylpentafluoropropionate, pentafluoropropionic acid methyl ester, propanoic acid, pentafluoro-, methyl ester, methyl pentafluoropropanoate, perfluoropropionic acid, methyl ester, pentafluoropropanoic acid methyl ester, 2,2,3,3,3-pentafluoro-propionic acid methyl ester, methylester kyseliny pentafluorpropionove
PubChem CID 9783
IUPAC Name methyl 2,2,3,3,3-pentafluoropropanoate
SMILES COC(=O)C(C(F)(F)F)(F)F

Specifications

Density 1.393
Boiling Point 59°C to 60°C
Flash Point −1°C (30°F)
Refractive Index 1.288
Quantity 5 g
UN Number UN3272
Beilstein 1777501
Formula Weight 178.06
Percent Purity 98%
Chemical Name or Material Methyl pentafluoropropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.