missing translation for 'onlineSavingsMsg'
Learn More

Methyl perfluorooctanoate, 97%, Thermo Scientific™

Product Code. 11406706
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406706 5 g 5g
11416706 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11406706 Supplier Thermo Scientific Alfa Aesar Supplier No. B23856.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376-27-2
Molecular Formula C9H3F15O2
Molecular Weight (g/mol) 428.097
MDL Number MFCD00039244
InChI Key XOCNYZFAMHDXJK-UHFFFAOYSA-N
Synonym methyl perfluorooctanoate, methyl pentadecafluorooctanoate, pentadecafluorooctanoic acid, methyl ester, methyl perfluorocaprylate, octanoic acid, pentadecafluoro-, methyl ester, perfluorooctanoic acid methyl ester, pentadecafluorooctanoic acid methyl ester, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, methyl ester, acmc-1cndk, methylpentadecafluorooctanoate
PubChem CID 67824
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
SMILES COC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.684
Boiling Point 158°C to 159°C
Flash Point 65°C (149°F)
Refractive Index 1.305
Quantity 5 g
Beilstein 1809817
Formula Weight 428.09
Percent Purity 97%
Chemical Name or Material Methyl perfluorooctanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.