missing translation for 'onlineSavingsMsg'
Learn More

Methyl Perfluorooctanoate, TRC

Product Code. 18082440
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18082440 1 g 1g
17917819 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18082440 Supplier Toronto Research Chemicals Supplier No. TRCM0102901G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376-27-2
Molecular Formula C9 H3 F15 O2
Molecular Weight (g/mol) 428.095
Synonym Perfluorooctanoic Acid-Methyl Ester, Methyl Pentadecafluorooctanoate, Octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, methyl ester, Octanoic acid, pentadecafluoro-, methyl ester (6CI,7CI,8CI,9CI), Methyl pentadecafluorooctanoate, Methyl perfluorooctanoate, P 1453, Perfluorooctanoic acid methyl ester
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
SMILES COC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H3F15O2/c1-26-2(25)3(10,11)4(12,13)5(14,15)6(16,17)7(18,19)8(20,21)9(22,23)24/h1H3
Formula Weight 427.989
Chemical Name or Material Perfluorooctanoic Acid- Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.