missing translation for 'onlineSavingsMsg'
Learn More

Methyl perfluorotetradecanoate, 96%, Thermo Scientific™

Product Code. 15489049
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15489049 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15489049 Supplier Thermo Scientific Alfa Aesar Supplier No. L16975.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifica

CAS 203302-99-2
Melting Point 95°C to 98°C
Molecular Formula C15H3F27O2
Linear Formula CF3(CF2)12CO2CH3
MDL Number MFCD00236620
Quantity 5 g
Synonym methyl perfluorotetradecanoate, tetradecanoic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluoro-,methyl ester, acmc-20akf3, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecanoic acid methyl ester, methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosakis fluoranyl tetradecanoate
InChI Key DRRCLKOIEBNFIE-UHFFFAOYSA-N
SMILES COC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecanoate
Molecular Weight (g/mol) 728.14
PubChem CID 2769340
Formula Weight 728.15
Percent Purity 96%
Chemical Name or Material Methyl perfluorotetradecanoate
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.