missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenylpropiolate, 97%

Product Code. 11490428
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490428 5 g 5g
11400438 25 g 25g
11410438 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490428 Supplier Thermo Scientific Alfa Aesar Supplier No. B24587.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4891-38-7 | C10H8O2 | 160.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4891-38-7
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00041685
InChI Key JFGWPXKGINUNDH-UHFFFAOYSA-N
Synonym methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate
PubChem CID 138378
IUPAC Name methyl 3-phenylprop-2-ynoate
SMILES COC(=O)C#CC1=CC=CC=C1

Specifications

Density 1.083
Boiling Point 109°C to 110°C (2 mmHg)
Flash Point >110°C (230°F)
Linear Formula C6H5C≡CCO2CH3
Refractive Index 1.562
Quantity 5 g
Beilstein 472813
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material Methyl phenylpropiolate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.