missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenylpropiolate, 98%

Product Code. 10061571 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10061571 5 g Glass bottle 5g
10564711 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10061571 Supplier Thermo Scientific Acros Supplier No. 334590050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4891-38-7 | C10H8O2 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4891-38-7
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00041685
InChI Key JFGWPXKGINUNDH-UHFFFAOYSA-N
Synonym methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate
PubChem CID 138378
IUPAC Name methyl 3-phenylprop-2-ynoate
SMILES COC(=O)C#CC1=CC=CC=C1

Specifications

Color Colorless to Yellow
Boiling Point 109°C to 110°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5620 to 1.564
Quantity 5 g
Formula Weight 160.17
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Methyl phenylpropiolate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.