missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl potassium malonate, 98%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Methyl potassium malonate is a protein advanced glycosylation inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl potassium malonate is a protein advanced glycosylation inhibitor.
Solubility
Soluble in methanol.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place.
Methyl potassium malonate is a protein advanced glycosylation inhibitor.
Solubility
Soluble in methanol.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
| Melting Point | ∽190°C (decomposition) |
| Color | White |
| Quantity | 25 g |
| Solubility Information | Soluble in methanol. |
| Formula Weight | 156.18 |
| Percent Purity | 98% |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl potassium malonate, Hygroscopic |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction