missing translation for 'onlineSavingsMsg'
Learn More

Methyl propiolate, 97%, Thermo Scientific™

Product Code. 10621765
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10621765 10 g Amber Glass Bottle 10g
10527664 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10621765 Supplier Thermo Scientific Maybridge Supplier No. SB01817EA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 922-67-8
Molecular Formula C4H4O2
Molecular Weight (g/mol) 84.074
MDL Number MFCD00008572
InChI Key IMAKHNTVDGLIRY-UHFFFAOYSA-N
Synonym methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester
PubChem CID 13536
IUPAC Name methyl prop-2-ynoate
SMILES COC(=O)C#C

Specifications

CAS Min % 97.0
Density 0.9400g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula HCidentical toCCO2CH3
Quantity 10 g
Formula Weight 84.07
Percent Purity 97%
Chemical Name or Material Methyl propiolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.