missing translation for 'onlineSavingsMsg'
Learn More

Methyl propiolate, 99%

Product Code. 10573731 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10573731 10 g Glass bottle 10g
10478320 50 g Glass bottle 50g
10699433 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10573731 Supplier Thermo Scientific Acros Supplier No. 174040100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 922-67-8 | C4H4O2 | 84.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 922-67-8
Molecular Formula C4H4O2
Molecular Weight (g/mol) 84.07
MDL Number MFCD00008572
InChI Key IMAKHNTVDGLIRY-UHFFFAOYSA-N
Synonym methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester
PubChem CID 13536
IUPAC Name methyl prop-2-ynoate
SMILES COC(=O)C#C

Specifications

Color Colorless
Density 0.9400g/mL
Boiling Point 98°C to 101°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HC∽CCO2CH3
Refractive Index 1.4070 to 1.409
Quantity 10 g
Beilstein 02, I, 208
Specific Gravity 0.94
Solubility Information Solubility in water: insoluble
Formula Weight 84.07
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl propiolate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl propiolate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.