missing translation for 'onlineSavingsMsg'
Learn More

Methyl propionate, 99%

Product Code. 11487127
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487127 2500 mL 2500mL
11467127 100 mL 100mL
11477127 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487127 Supplier Thermo Scientific Alfa Aesar Supplier No. A14944.0F
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 554-12-1 | C4H8O2 | 88.106 g/mol

It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.

Solubility
Soluble in water (5g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-12-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009306
InChI Key RJUFJBKOKNCXHH-UHFFFAOYSA-N
Synonym methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural
PubChem CID 11124
IUPAC Name methyl propanoate
SMILES CCC(=O)OC

Specifications

Melting Point -88°C
Density 0.915
Boiling Point 78°C to 80°C
Flash Point −2°C (28°F)
Assay Percent Range 99%
Refractive Index 1.377
Quantity 2500 mL
UN Number UN1248
Beilstein 1737628
Merck Index 14,6112
Solubility Information Soluble in water (5g/L).
Formula Weight 88.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl propionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.