missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Methyl protodioscin

Product Code. 16439148
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439148 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439148 Supplier MedChemExpress Supplier No. HYN0863

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Methyl protodioscin(NSC-698790) is a furostanol bisglycoside with antitumor properties; shows to reduce proliferation, cause cell cycle arrest.

Specifications

Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Molecular Weight (g/mol) 1063.23 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 54522-52-0
Product Type Natural Products
Purity 98.0%
Solubility DMSO : 140 mg/mL (131.67 mM∣Need ultrasonic) ∣H2O : 25 mg/mL (23.51 mM∣Need ultrasonic)
Synonym NSC-698790∣ Smilax saponin B
Target Apoptosis
Quantity 10 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.