missing translation for 'onlineSavingsMsg'
Learn More

Methyl pyrazine-2-carboxylate, 98+%, Thermo Scientific™

Product Code. 11479892
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479892 100 g 100g
11459892 5 g 5g
11469892 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479892 Supplier Thermo Scientific Alfa Aesar Supplier No. A12028.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6164-79-0
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00014611
InChI Key TWIIRMSFZNYMQE-UHFFFAOYSA-N
Synonym methyl 2-pyrazinecarboxylate, methyl pyrazinoate, pyrazinoic acid methyl ester, methylpyrazine-2-carboxylate, pyrazinecarboxylic acid, methyl ester, methyl pyrazinecarboxylate, 2-pyrazinecarboxylic acid, methyl ester, pyrazine-2-carboxylic acid, methyl ester, 2-methylpyrazine carboxylate, pyrazine-2-carboxylic acid methyl ester
PubChem CID 72662
IUPAC Name methyl pyrazine-2-carboxylate
SMILES COC(=O)C1=NC=CN=C1

Specifications

Melting Point 56°C to 60°C
Flash Point >100°C (212°F)
Quantity 100 g
Beilstein 118345
Merck Index 14,7959
Formula Weight 138.13
Percent Purity ≥98%
Chemical Name or Material Methyl pyrazine-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.