missing translation for 'onlineSavingsMsg'
Learn More

Methyl pyruvate, 94%

Product Code. 10503254 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10503254 25 g Glass bottle 25g
10081330 100 g Glass bottle 100g
10710585 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10503254 Supplier Thermo Scientific Acros Supplier No. 127650250
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 600-22-6 | C4H6O3 | 102.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 600-22-6
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00008754
InChI Key CWKLZLBVOJRSOM-UHFFFAOYSA-N
Synonym methyl pyruvate, pyruvic acid, methyl ester, pyruvic acid methyl ester, methyl 2-oxopropionate, propanoic acid, 2-oxo-, methyl ester, methyl-pyruvate, methylglyoxylic acid methyl ester, unii-3kjm65g5xl, 3kjm65g5xl, 2-oxo-propionic acid methyl ester
PubChem CID 11748
ChEBI CHEBI:51850
IUPAC Name methyl 2-oxopropanoate
SMILES CC(=O)C(=O)OC

Specifications

Color Colorless to Yellow
Density 1.1300g/mL
Boiling Point 134.0°C to 137.0°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Linear Formula CH3COCO2CH3
Refractive Index 1.4065
Quantity 25 g
Beilstein 03, 616
Merck Index 15, 8135
Specific Gravity 1.13
Solubility Information Solubility in water: soluble.
Formula Weight 102.09
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Methyl pyruvate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.